siRNA Details
| Gene Symbol: LOC101928617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135036.1 | 3 | 1421 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR622414.1 | - | 3 | 1325 | Chr. 12 - 92146181 - 92185784 [+] on Build GRCh38 | |

LOC101928617
Genomic Map