siRNA Details
| Gene Symbol: G3BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006714749.3 | XP_006714812.1 | 12 | 8361 | ||
| XM_006714750.3 | XP_006714813.1 | 12 | 8300 | |||
| XM_017008922.1 | XP_016864411.1 | 12 | 8647 | |||
| XM_017008923.1 | XP_016864412.1 | 12 | 8616 | |||
| GenBank mRNA | AK128316.1 | - | 1 | 4138 | ||

G3BP1
Genomic Map