siRNA Details
| Gene Symbol: KLHL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022805.1 | - | 1 | 3958 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | N99347.1 | - | 2 | 1537 | Chr. 9 - 21324052 - 21326339 [-] on Build GRCh38 | |

KLHL9
Genomic Map