siRNA Details
| Gene Symbol: HEATR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005257570.3 | XP_005257627.1 | 21 | 5175 | ||
| XM_011525110.2 | XP_011523412.1 | 19 | 4874 | |||
| XM_011525111.2 | XP_011523413.1 | 19 | 4836 | |||
| XM_017024936.1 | XP_016880425.1 | 20 | 5248 | |||
| XM_017024937.1 | XP_016880426.1 | 20 | 6924 | |||
| XM_017024938.1 | XP_016880427.1 | 20 | 4980 | |||
| XM_017024939.1 | XP_016880428.1 | 20 | 5251 | |||
| GenBank mRNA | BC040714.1 | - | 1 | 852 | ||

HEATR6
Genomic Map