siRNA Details
| Gene Symbol: NAV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ488202.1 | - | 1 | 51 | ||
| Gene Symbol: NAV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020166.1 | XP_016875655.1 | 1 | 330 | ||
| XM_017020167.1 | XP_016875656.1 | 1 | 330 | |||

NAV2
Genomic Map