siRNA Details
| Gene Symbol: LINC00511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033876.1 | 4 | 471 | |||
| GenBank mRNA | AK129994.1 | - | 4 | 471 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000581549.1 | - | 4 | 471 | Chr. 17 - 72598041 - 72640472 [-] on Build GRCh38 | |

LINC00511
Genomic Map