siRNA Details
| Gene Symbol: ZNRF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046851.1 | 3 | 742 | |||
| GenBank mRNA | AK095488.1 | - | 3 | 741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRF3-AS1| Gene Symbol: ZNRF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046851.1 | 3 | 742 | |||
| GenBank mRNA | AK095488.1 | - | 3 | 741 | ||