siRNA Details
| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127059.1 | - | 1 | 3168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA430431.1 | - | 1 | 209 | Chr. 21 - 44936303 - 44936938 [-] on Build GRCh38 | |

LINC01547
Genomic Map