siRNA Details
| Gene Symbol: STUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003665.3 | NP_001003665.1 | 4 | 5619 | ||
| XM_011544183.2 | XP_011542485.1 | 3 | 6172 | |||
| GenBank mRNA | AF035308.1 | - | 1 | 1167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STUM| Gene Symbol: STUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003665.3 | NP_001003665.1 | 4 | 5619 | ||
| XM_011544183.2 | XP_011542485.1 | 3 | 6172 | |||
| GenBank mRNA | AF035308.1 | - | 1 | 1167 | ||