siRNA Details
| Gene Symbol: STUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003665.3 | NP_001003665.1 | 4 | 5366 | ||
| XM_011544183.2 | XP_011542485.1 | 3 | 5919 | |||
| GenBank mRNA | AF035308.1 | - | 1 | 914 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI914560.1 | - | 1 | 13 | Chr. 1 - 226606863 - 226607766 [+] on Build GRCh38 | |

STUM
Genomic Map