siRNA Details
| Gene Symbol: DLEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC033120.1 | - | 1 | 594 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM423396.1 | - | 1 | 594 | Chr. 13 - 50040088 - 50041038 [-] on Build GRCh38 | |

DLEU2
Genomic Map