siRNA Details
| Gene Symbol: CEP85L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042475.2 | NP_001035940.1 | 13 | 6849 | ||
| NM_001178035.1 | NP_001171506.1 | 14 | 6337 | |||
| XM_005266970.1 | XP_005267027.1 | 13 | 6369 | |||
| XM_005266971.1 | XP_005267028.1 | 13 | 6332 | |||
| XM_005266972.4 | XP_005267029.1 | 11 | 6472 | |||
| XM_006715475.3 | XP_006715538.1 | 14 | 6545 | |||
| XM_011535809.2 | XP_011534111.1 | 14 | 7440 | |||
| XM_011535810.2 | XP_011534112.1 | 14 | 6294 | |||
| XM_011535811.1 | XP_011534113.1 | 13 | 6235 | |||
| XM_017010846.1 | XP_016866335.1 | 15 | 6473 | |||
| XM_017010847.1 | XP_016866336.1 | 12 | 6484 | |||
| GenBank mRNA | AL133101.1 | - | 1 | 1910 | ||
| BC045657.1 | - | 1 | 3435 | |||

CEP85L
Genomic Map