siRNA Details
| Gene Symbol: LOC101927907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110284.1 | 3 | 194 | |||
| GenBank mRNA | BC018017.1 | - | 3 | 194 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF700684.1 | - | 195 | Chr. - - [-] on Build | ||

LOC101927907
Genomic Map