siRNA Details
| Gene Symbol: LOC101929470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125817.1 | 3 | 989 | |||
| GenBank mRNA | AK054638.1 | - | 3 | 989 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000507496.5 | - | 3 | 989 | Chr. 8 - 29815004 - 29828504 [-] on Build GRCh38 | |

LOC101929470
Genomic Map