siRNA Details
| Gene Symbol: LOC101929470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125817.1 | 3 | 907 | |||
| GenBank mRNA | AK054638.1 | - | 3 | 907 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929470| Gene Symbol: LOC101929470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125817.1 | 3 | 907 | |||
| GenBank mRNA | AK054638.1 | - | 3 | 907 | ||