siRNA Details
| Gene Symbol: LOC100996681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034565.1 | - | 2 | 934 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589967.1 | - | 2 | 934 | Chr. 19 - 33555568 - 33557470 [+] on Build GRCh38 | |

LOC100996681
Genomic Map