siRNA Details
Gene Symbol: AVPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016494.1 | XP_016871983.1 | 1 | 145 | ||
GenBank mRNA | AY505173.1 | - | 1 | 117 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AVPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016494.1 | XP_016871983.1 | 1 | 145 | ||
GenBank mRNA | AY505173.1 | - | 1 | 117 |