siRNA Details
Gene Symbol: LINC01562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127900.1 | - | 2 | 2197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000366181.2 | - | 2192 | Chr. 5 - 176376243 - 176376645 [+] on Build GRCh38 |
