siRNA Details
| Gene Symbol: LINC01562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127900.1 | - | 2 | 387 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA927036.1 | - | 2 | 387 | Chr. 1 - 51196879 - 51235096 [-] on Build GRCh38 | |

LINC01562
Genomic Map