siRNA Details
| Gene Symbol: LOC101928372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110695.1 | 5, 6 | 859 | |||
| GenBank mRNA | BC035240.1 | - | 5, 6 | 859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928372| Gene Symbol: LOC101928372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110695.1 | 5, 6 | 859 | |||
| GenBank mRNA | BC035240.1 | - | 5, 6 | 859 | ||