siRNA Details
| Gene Symbol: LOC101241902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077236.1 | 1 | 1010 | |||
| GenBank mRNA | BC072451.1 | - | 1 | 1010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101241902| Gene Symbol: LOC101241902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077236.1 | 1 | 1010 | |||
| GenBank mRNA | BC072451.1 | - | 1 | 1010 | ||