siRNA Details
| Gene Symbol: LOC101241902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077236.1 | 1 | 20 | |||
| GenBank mRNA | BC072451.1 | - | 1 | 20 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ883439.1 | - | 1 | 19 | Chr. 8 - 78723553 - 78724373 [-] on Build GRCh38 | |

LOC101241902
Genomic Map