siRNA Details
Gene Symbol: LOC286068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096200.1 | - | 1 | 742 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC015720.1 | - | 1 | 1122 | Chr. 8 - 48428273 - 48431041 [-] on Build GRCh38 |
