siRNA Details
| Gene Symbol: LEPROT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094801.1 | - | 1 | 1421 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125268.1 | - | 1 | 850 | Chr. 1 - 16464556 - 16466824 [-] on Build GRCh38 | |

LEPROT
Genomic Map