siRNA Details
| Gene Symbol: LOC284648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036490.1 | 1 | 287 | |||
| GenBank mRNA | AW297673.1 | - | 1 | 287 | ||
| BC034591.1 | - | 1 | 173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284648| Gene Symbol: LOC284648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036490.1 | 1 | 287 | |||
| GenBank mRNA | AW297673.1 | - | 1 | 287 | ||
| BC034591.1 | - | 1 | 173 | |||