siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF319522.1 | - | 3, 4 | 287 | ||
| U81001.1 | - | LIT2197 IC-SNURF-SNRPN 3 prime noncoding transcripts |
1 | 964 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB033107.1 | - | 3, 4 | 374 | Chr. 15 - 24954948 - 24980058 [+] on Build GRCh38 | |

SNRPN
Genomic Map