siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125976.1 | 5 | 880 | |||
| GenBank mRNA | BC034623.1 | - | 5 | 546 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG772721.1 | - | 5 | 551 | Chr. 1 - 119140726 - 119233856 [+] on Build GRCh38 | |

LOC101929147
Genomic Map