siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125976.1 | 5 | 1434 | |||
| GenBank mRNA | BC034623.1 | - | 5 | 1100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929147| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125976.1 | 5 | 1434 | |||
| GenBank mRNA | BC034623.1 | - | 5 | 1100 | ||