siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125975.1 | 2, 3 | 471 | |||
| NR_125976.1 | 2, 3 | 471 | ||||
| GenBank mRNA | AK125759.1 | - | 2, 3 | 157 | ||
| BC034623.1 | - | 2, 3 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929147| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125975.1 | 2, 3 | 471 | |||
| NR_125976.1 | 2, 3 | 471 | ||||
| GenBank mRNA | AK125759.1 | - | 2, 3 | 157 | ||
| BC034623.1 | - | 2, 3 | 137 | |||