siRNA Details
Gene Symbol: LOC101929147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125976.1 | 5 | 824 | |||
GenBank mRNA | BC034623.1 | - | 5 | 490 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125976.1 | 5 | 824 | |||
GenBank mRNA | BC034623.1 | - | 5 | 490 |