siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125975.1 | 3 | 516 | |||
| NR_125976.1 | 3 | 516 | ||||
| NR_125977.1 | 2 | 449 | ||||
| GenBank mRNA | AK125759.1 | - | 3 | 202 | ||
| BC034623.1 | - | 3 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929147| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125975.1 | 3 | 516 | |||
| NR_125976.1 | 3 | 516 | ||||
| NR_125977.1 | 2 | 449 | ||||
| GenBank mRNA | AK125759.1 | - | 3 | 202 | ||
| BC034623.1 | - | 3 | 182 | |||