siRNA Details
| Gene Symbol: PRICKLE2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046702.1 | 4 | 2971 | |||
| GenBank mRNA | AK056491.1 | - | 4 | 2969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRICKLE2-AS3| Gene Symbol: PRICKLE2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046702.1 | 4 | 2971 | |||
| GenBank mRNA | AK056491.1 | - | 4 | 2969 | ||