siRNA Details
| Gene Symbol: PRICKLE2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046702.1 | 2 | 84 | |||
| GenBank mRNA | AK056491.1 | - | 2 | 84 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA742626.1 | - | 2 | 84 | Chr. 3 - 64187544 - 64199035 [+] on Build GRCh38 | |

PRICKLE2-AS3
Genomic Map