siRNA Details
| Gene Symbol: SCML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094942.1 | - | 1 | 231 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA284137.1 | - | 1 | 231 | Chr. X - 17737848 - 17738430 [+] on Build GRCh38 | |

SCML1
Genomic Map