siRNA Details
| Gene Symbol: SLC26A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF161369.1 | - | 1 | 517 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF150415.1 | - | 1 | 517 | Chr. 19 - 15273835 - 15274702 [-] on Build GRCh38 | |

SLC26A6
Genomic Map