siRNA Details
| Gene Symbol: LINGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258282.1 | NP_001245211.1 | 7 | 3121 | ||
| NM_152570.2 | NP_689783.1 | 7 | 3201 | |||
| XM_011517719.1 | XP_011516021.1 | 6 | 4069 | |||
| XM_011517724.2 | XP_011516026.1 | 5 | 7088 | |||
| XM_011517728.2 | XP_011516030.1 | 2 | 6867 | |||
| XM_017014303.1 | XP_016869792.1 | 11 | 6972 | |||
| XM_017014304.1 | XP_016869793.1 | 4 | 9492 | |||
| XM_017014305.1 | XP_016869794.1 | 5 | 9075 | |||
| XM_017014306.1 | XP_016869795.1 | 3 | 6236 | |||
| XM_017014307.1 | XP_016869796.1 | 2 | 3709 | |||

LINGO2
Genomic Map