siRNA Details
| Gene Symbol: LINGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011517719.1 | XP_011516021.1 | 6 | 4101 | ||
| XM_011517724.2 | XP_011516026.1 | 5 | 7120 | |||
| XM_011517728.2 | XP_011516030.1 | 2 | 6899 | |||
| XM_017014303.1 | XP_016869792.1 | 11 | 7004 | |||
| XM_017014304.1 | XP_016869793.1 | 4 | 9524 | |||
| XM_017014305.1 | XP_016869794.1 | 5 | 9107 | |||
| XM_017014306.1 | XP_016869795.1 | 3 | 6268 | |||
| XM_017014307.1 | XP_016869796.1 | 2 | 3741 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR618670.1 | - | 1 | 660 | Chr. 9 - 27948078 - 27948748 [-] on Build GRCh38 | |

LINGO2
Genomic Map