siRNA Details
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017872.1 | XP_016873361.1 | 1 | 463 | ||
GenBank mRNA | S72370.1 | - | 1 | 31 | ||
U30890.1 | - | 1 | 80 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017872.1 | XP_016873361.1 | 1 | 463 | ||
GenBank mRNA | S72370.1 | - | 1 | 31 | ||
U30890.1 | - | 1 | 80 |