siRNA Details
| Gene Symbol: LOC100506688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104614.1 | 2 | 992 | |||
| NR_104615.1 | 2 | 926 | ||||
| GenBank mRNA | AK126199.1 | - | 2 | 960 | ||
| AK127451.1 | - | 1 | 126 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA394528.1 | - | 1 | 126 | Chr. 5 - 989241 - 989827 [-] on Build GRCh38 | |

LOC100506688
Genomic Map