siRNA Details
| Gene Symbol: LOC100506688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104614.1 | 2 | 2111 | |||
| NR_104615.1 | 2 | 2045 | ||||
| GenBank mRNA | AK126199.1 | - | 2 | 2079 | ||
| AK127451.1 | - | 1 | 1245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506688| Gene Symbol: LOC100506688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104614.1 | 2 | 2111 | |||
| NR_104615.1 | 2 | 2045 | ||||
| GenBank mRNA | AK126199.1 | - | 2 | 2079 | ||
| AK127451.1 | - | 1 | 1245 | |||