siRNA Details
| Gene Symbol: LOC84214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136837.1 | - | 1 | 2215 | ||
| Gene Symbol: LOC105376666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017018640.1 | XP_016874129.1 | 1 | 5534 | ||

LOC84214
Genomic Map