siRNA Details
| Gene Symbol: LOC105374181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130735.1 | - | 1 | 1732 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA675109.1 | - | 1 | 465 | Chr. 3 - 158871162 - 158871745 [+] on Build GRCh38 | |

LOC105374181
Genomic Map