siRNA Details
| Gene Symbol: LOC105374181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130735.1 | - | 1948 | |||
| Gene Symbol: TRIM56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516589.2 | XP_011514891.1 | 2 | 6340 | ||

LOC105374181
Genomic Map