siRNA Details
| Gene Symbol: GCRG224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF438406.1 | - | 1 | 713 | ||
| Gene Symbol: PRCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW627417.1 | - | 1 | 144 | ||

GCRG224
Genomic Map