siRNA Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278656.1 | NP_001265585.1 | 4 | 3108 | ||
| NM_004175.4 | NP_004166.1 | 4 | 3122 | |||
| NR_103819.1 | 3 | 2929 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNRPD3| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278656.1 | NP_001265585.1 | 4 | 3108 | ||
| NM_004175.4 | NP_004166.1 | 4 | 3122 | |||
| NR_103819.1 | 3 | 2929 | ||||