siRNA Details
| Gene Symbol: PPIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005246966.1 | XP_005247023.1 | 14 | 6222 | ||
| XM_005246967.1 | XP_005247024.1 | 14 | 6269 | |||
| GenBank mRNA | AK124958.1 | - | 1 | 1822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPIG| Gene Symbol: PPIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005246966.1 | XP_005247023.1 | 14 | 6222 | ||
| XM_005246967.1 | XP_005247024.1 | 14 | 6269 | |||
| GenBank mRNA | AK124958.1 | - | 1 | 1822 | ||