siRNA Details
| Gene Symbol: ZNF75D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110381.1 | 3, 4 | 1294 | |||
| GenBank mRNA | AY730281.1 | - | 2, 3 | 488 | ||
| BC029787.1 | - | 3, 4 | 1294 | |||
| BG186944.1 | - | 2, 3 | 313 | |||
| Gene Symbol: LOC100287728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103770.1 | 1, 2 | 50 | |||

ZNF75D
Genomic Map