siRNA Details
| Gene Symbol: ZNF75D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110381.1 | 3 | 1242 | |||
| GenBank mRNA | AY730281.1 | - | 2 | 436 | ||
| BC029787.1 | - | 3 | 1242 | |||
| BG186944.1 | - | 2 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF75D| Gene Symbol: ZNF75D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110381.1 | 3 | 1242 | |||
| GenBank mRNA | AY730281.1 | - | 2 | 436 | ||
| BC029787.1 | - | 3 | 1242 | |||
| BG186944.1 | - | 2 | 261 | |||