siRNA Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020980.1 | XP_016876469.1 | 4 | 1528 | ||
| XM_017020981.1 | XP_016876470.1 | 3 | 1052 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK094455.1 | - | 661 | Chr. - - [-] on Build | ||

SLC25A29
Genomic Map