siRNA Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020980.1 | XP_016876469.1 | 4 | 1774 | ||
| XM_017020981.1 | XP_016876470.1 | 3 | 1298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A29| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020980.1 | XP_016876469.1 | 4 | 1774 | ||
| XM_017020981.1 | XP_016876470.1 | 3 | 1298 | |||